EMAIL THIS PAGE TO A FRIEND
74902 Sigma-Aldrich

1-Octene

purum, ≥97.0% (GC)

  • CAS Number 111-66-0

  • Linear Formula CH3(CH2)5CH=CH2

  • Molecular Weight 112.21

  •  Beilstein Registry Number 1734497

  •  EC Number 203-893-7

  •  MDL number MFCD00009548

Purchase

Properties

Related Categories Acyclic, Alkenes, Building Blocks, Chemical Synthesis, Organic Building Blocks More...
vapor density   3.9 (vs air)
vapor pressure   36 mmHg ( 38 °C)
InChI Key   KWKAKUADMBZCLK-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
form   liquid
autoignition temp.   446 °F
expl. lim.   3.9 %
refractive index   n20/D 1.408(lit.)
  n20/D 1.409(lit.)
  n20/D 1.409
bp   122-123 °C(lit.)
mp   −101 °C(lit.)
density   0.715 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN3295 - class 3 - PG 2 - EHS - VP <= 110kPa - Hydrocarbons, liquid, n.o.s., HI: A, S (not BR)
WGK Germany 
2
Flash Point(F) 
50 °F
Flash Point(C) 
10 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

O4806 1-Octene, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?