EMAIL THIS PAGE TO A FRIEND
74990 Sigma-Aldrich

Octylamine

puriss., ≥99.0% (GC)

Synonym: 1-Aminooctane, n-Octylamine, Caprylamine

  • CAS Number 111-86-4

  • Linear Formula CH3(CH2)7NH2

  • Molecular Weight 129.24

  •  Beilstein Registry Number 1679227

  •  EC Number 203-916-0

  •  MDL number MFCD00008247

  •  PubChem Substance ID 329766027

Purchase

Properties

vapor pressure   1 mmHg ( 20 °C)
InChI Key   IOQPZZOEVPZRBK-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (GC)
refractive index   n20/D 1.429(lit.)
  n20/D 1.430
bp   175-177 °C(lit.)
mp   −5-−1 °C(lit.)
density   0.782 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
RIDADR 
UN2734 - class 8 - PG 1 - EHS - Amines, liquid, corrosive, flammable, n.o.s., HI: all
WGK Germany 
2
RTECS 
RG8050000
Flash Point(F) 
140 °F
Flash Point(C) 
60 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

O5802 Octylamine, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?