EMAIL THIS PAGE TO A FRIEND
75010 Sigma-Aldrich

1-Phenyloctane

purum, ≥98.0% (GC)

Synonym: Octylbenzene

Purchase

Properties

Related Categories Arenes, Building Blocks, Chemical Synthesis, Organic Building Blocks
InChI Key   CDKDZKXSXLNROY-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.484(lit.)
  n20/D 1.484
bp   261-263 °C(lit.)
mp   −36 °C(lit.)
density   0.858 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
224.6 °F
Flash Point(C) 
107 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

113190 1-Phenyloctane, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?