• USA Home
  • 75079 - Octyl α-D-glucopyranoside

EMAIL THIS PAGE TO A FRIEND
75079 Sigma-Aldrich

Octyl α-D-glucopyranoside

≥98.0% (TLC)

Synonym: n-Octyl α-glucoside

  • CAS Number 29781-80-4

  • Empirical Formula (Hill Notation) C14H28O6

  • Molecular Weight 292.37

  •  Beilstein Registry Number 4232620

  •  MDL number MFCD00070008

Purchase

Description

Other Notes

Typical amphiphile, suitable for the crystallization of membrane proteins

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

O0630 Octyl α-D-glucopyranoside, ≥98% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?