EMAIL THIS PAGE TO A FRIEND
75093 Sigma-Aldrich

Oleic acid

from suet, natural, ≥60% (GC)

Synonym: cis-9-Octadecenoic acid, Elainic acid

  • CAS Number 112-80-1

  • Linear Formula CH3(CH2)7CH=CH(CH2)7COOH

  • Molecular Weight 282.46

  •  Beilstein Registry Number 1726542

  •  EC Number 204-007-1

  •  MDL number MFCD00064242

Purchase

Properties

Related Categories Building Blocks, C13 to C42+, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
biological source   from suet
vapor pressure   1 mmHg ( 176 °C)
InChI Key   ZQPPMHVWECSIRJ-KTKRTIGZSA-N
assay   ≥60% (GC)
quality   natural
refractive index   n20/D 1.459(lit.)
  n20/D 1.459
bp   194-195 °C/1.2 mmHg(lit.)
mp   13-14 °C(lit.)
density   0.89 g/mL at 25 °C(lit.)

Description

Biochem/physiol Actions

Activates protein kinase C in hepatocytes.

Uncouples oxidative phosphorylation. Inhibits 2,4-dinitrophenol−stimulated ATPase. Action reversed by adding serum albumin.

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
WGK Germany 
1
RTECS 
RG2275000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

364525 Oleic acid, technical grade, 90%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?