EMAIL THIS PAGE TO A FRIEND
75165 Sigma-Aldrich

Sodium oleate

≥98.5% (GC)

Synonym: cis-9-Octadecenoic acid sodium salt, Oleic acid sodium salt

  • CAS Number 143-19-1

  • Linear Formula CH3(CH2)7CH=CH(CH2)7COONa

  • Molecular Weight 304.44

  •  Beilstein Registry Number 4046357

  •  EC Number 205-591-0

  •  MDL number MFCD00004438

  •  PubChem Substance ID 329766228

Purchase

Properties

Related Categories Biochemicals and Reagents, Fatty Acids and conjugates, Fatty Acyls, Lipids, Monoenoic fatty acids,
InChI Key   BCKXLBQYZLBQEK-KVVVOXFISA-M
assay   ≥98.5% (GC)
impurities   ≤5% water
mp   232-235 °C(lit.)
solubility   methanol: soluble50 mg/mL, clear, colorless
storage temp.   −20°C

Description

Other Notes

Potent non-phagocytic stimulator of cyanide-resistant oxidative metabolism

Application

Oleic acid is used to investigate effects on apolipoprotein B synthesis, degradation and secretion in cultured cells.

Biochem/physiol Actions

Oleic acid increases hepatic secretion of apolipoprotein B100 in hepatocyte cell lines and in mice . Oleic acid inhibits apolipoprotein B100 secretion at higher physiologic doses .

Price and Availability


Laboratory Gloves

The Future of Bioimaging
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
RK1200000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?