EMAIL THIS PAGE TO A FRIEND
75200 Sigma-Aldrich

Ethyl heptanoate

purum, ≥96.0% (GC)

Synonym: Ethyl enanthate

  • CAS Number 106-30-9

  • Linear Formula CH3(CH2)5COOC2H5

  • Molecular Weight 158.24

  •  Beilstein Registry Number 1752311

  •  EC Number 203-382-9

  •  MDL number MFCD00009538

  •  PubChem Substance ID 24886804

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, C8 to C9, Carbonyl Compounds, Chemical Synthesis,
InChI Key   TVQGDYNRXLTQAP-UHFFFAOYSA-N
grade   purum
assay   ≥96.0% (GC)
refractive index   n20/D 1.412(lit.)
  n20/D 1.413
bp   188-189 °C(lit.)
mp   −66 °C(lit.)
density   0.87 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
1
RTECS 
MJ2087000
Flash Point(F) 
165.2 °F
Flash Point(C) 
74 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

112364 Ethyl heptanoate, ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?