EMAIL THIS PAGE TO A FRIEND
75342 Sigma-Aldrich

Oleyl alcohol

technical, ~60% (GC)

Synonym: Z-9-Dodecen-1-ol, cis-9-Octadecen-1-ol

  • CAS Number 143-28-2

  • Linear Formula CH3(CH2)7CH=CH(CH2)7CH2OH

  • Molecular Weight 268.48

  •  Beilstein Registry Number 1723962

  •  EC Number 205-597-3

  •  MDL number MFCD00002993

  •  PubChem Substance ID 329766362

Purchase

Properties

Related Categories Acyclic, Alkenes, Biochemicals and Reagents, Building Blocks, Chemical Synthesis,
InChI Key   ALSTYHKOOCGGFT-KTKRTIGZSA-N
grade   technical
assay   ~60% (GC)
refractive index   n20/D 1.46(lit.)
  n20/D 1.460
bp   207 °C/13 mmHg(lit.)
mp   0-5.0 °C(lit.)
iodine value   ~95
density   0.849 g/mL at 25 °C(lit.)

Price and Availability


The Future of Bioimaging
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
RG4120000
Flash Point(F) 
338 °F
Flash Point(C) 
170 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

369314 Oleyl alcohol, technical grade, 85%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?