• USA Home
  • 75379 - Lewatit® VP OC 1064 MD PH

EMAIL THIS PAGE TO A FRIEND
75379 Sigma-Aldrich

Lewatit® VP OC 1064 MD PH

adsorption resin crosslinked polystyrene matrix with a very high surface area without any functional group

Purchase

Properties

Related Categories Adsorbents (Resins), Analytical/Chromatography, LPLC Separation Media, LPLC/TLC/Paper Chromatography, Miscellaneous More...
loss   ~55% loss on drying, 110 °C
particle size   0.4-1 mm

Description

Application

Especially suited for isolation/purification of natural or synthetic products in the chemical industry. Can be used to remove chlorinated and nitrated hydrocarbons, phenols, aromatic compounds, pestizides, detergents, colored compounds or other organic impurities from waste water.

Legal Information

Lewatit is a registered trademark of Lanxess Deutschland GmbH

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?