• USA Home
  • 75510 - Orotic acid monohydrate

EMAIL THIS PAGE TO A FRIEND
75510 Sigma-Aldrich

Orotic acid monohydrate

purum, ≥97.0% (T)

Synonym: 2,6-Dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid

  • CAS Number 50887-69-9

  • Empirical Formula (Hill Notation) C5H4N2O4 · H2O

  • Molecular Weight 174.11

  •  Beilstein Registry Number 7604036

  •  MDL number MFCD00149398

  •  PubChem Substance ID 329766478

Purchase

Properties

Related Categories Building Blocks, C4 to C5, Cell Biology, Chemical Synthesis, Heterocyclic Building Blocks,
InChI Key   YXUZGLGRBBHYFZ-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (T)
ign. residue   ≤0.05%
mp   >300 °C(lit.)

Price and Availability


Laboratory Gloves

All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

O8402 Orotic acid monohydrate, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?