• USA Home
  • 75536 - Phosphate ionophore II

EMAIL THIS PAGE TO A FRIEND
75536 Sigma-Aldrich

Phosphate ionophore II

Selectophore

  • Empirical Formula (Hill Notation) C40H50FeN2O4

  • Molecular Weight 678.68

Purchase

Properties

grade   Selectophore

Description

Legal Information

Selectophore is a trademark of Sigma-Aldrich Chemie GmbH

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?