• USA Home
  • 75585 - Tetraethyl orthocarbonate

EMAIL THIS PAGE TO A FRIEND
75585 Sigma-Aldrich

Tetraethyl orthocarbonate

purum, ≥97.0% (GC)

  • CAS Number 78-09-1

  • Linear Formula C(OC2H5)4

  • Molecular Weight 192.25

  •  Beilstein Registry Number 1747401

  •  EC Number 201-082-2

  •  MDL number MFCD00009221

  •  PubChem Substance ID 329766145

Purchase

Properties

Related Categories Acetals/Ketals/Ortho Esters, Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds More...
InChI Key   CWLNAJYDRSIKJS-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
impurities   ≤0.05% water
refractive index   n20/D 1.392(lit.)
  n20/D 1.392
bp   159 °C(lit.)
density   0.915 g/mL at 20 °C
  0.919 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Review

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 3272 3 / PGIII
WGK Germany 
3
Flash Point(F) 
127.4 °F
Flash Point(C) 
53 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

163627 Tetraethyl orthocarbonate, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?