• USA Home
  • 75588 - Tetramethyl orthocarbonate

EMAIL THIS PAGE TO A FRIEND
75588 Sigma-Aldrich

Tetramethyl orthocarbonate

purum, ≥97.0% (GC)

  • CAS Number 1850-14-2

  • Linear Formula C(OCH3)4

  • Molecular Weight 136.15

  •  Beilstein Registry Number 1737682

  •  EC Number 217-438-5

  •  MDL number MFCD00008473

Purchase

Properties

Related Categories Acetals/Ketals/Ortho Esters, Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds More...
InChI Key   AHJWSRRHTXRLAQ-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.385(lit.)
  n20/D 1.385
bp   114 °C(lit.)
mp   −5 °C(lit.)
density   1.023 g/mL at 25 °C(lit.)

Description

Other Notes

Review

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3272 3 / PGII
WGK Germany 
3
Flash Point(F) 
71.6 °F
Flash Point(C) 
22 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

132624 Tetramethyl orthocarbonate, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?