• USA Home
  • 75601 - Triethyl orthopropionate

EMAIL THIS PAGE TO A FRIEND
75601 Sigma-Aldrich

Triethyl orthopropionate

purum, ≥97.0% (GC)

  • CAS Number 115-80-0

  • Linear Formula CH3CH2C(OC2H5)3

  • Molecular Weight 176.25

  •  Beilstein Registry Number 906798

  •  EC Number 204-108-0

  •  MDL number MFCD00009226

  •  PubChem Substance ID 329766156

Purchase

Properties

Related Categories Acetals/Ketals/Ortho Esters, Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds More...
InChI Key   FGWYWKIOMUZSQF-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.402(lit.)
  n20/D 1.402
bp   155-160 °C(lit.)
density   0.876 g/mL at 25 °C(lit.)

Description

Caution

may discolor to yellow on storage

Other Notes

Review of its use

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3272 3 / PGIII
WGK Germany 
1
Flash Point(F) 
140 °F
Flash Point(C) 
60 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

T60607 Triethyl orthopropionate, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?