• USA Home
  • 75612 - Osmium standard for AAS

EMAIL THIS PAGE TO A FRIEND
75612 Sigma-Aldrich

Osmium standard for AAS

ready-to-use, in hydrochloric acid

Synonym: Ammonium hexachloroosmate(IV) solution

Purchase

Properties

InChI Key   SYQBFIAQOQZEGI-UHFFFAOYSA-N
shelf life   limited shelf life, expiry date on the label
concentration   in hydrochloric acid
  1.000 g/L Os
application(s)   ICP/AAS: suitable
format   single component solution

Description

Preparation Note

prepared with (NH4)2[OsCl6] and HCl (~2 M)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
UN 1789 8 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?