EMAIL THIS PAGE TO A FRIEND
75690 Sigma-Aldrich

Oxalic acid

purum, anhydrous, ≥97.0% (RT)

Purchase

Properties

Related Categories Building Blocks, C1 to C5, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
vapor density   4.4 (vs air)
vapor pressure   <0.01 mmHg ( 20 °C)
InChI Key   MUBZPKHOEPUJKR-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (RT)
impurities   ~1% water
mp   189.5 °C (dec.)(lit.)
  189.5 °C (dec.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
RO2450000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?