EMAIL THIS PAGE TO A FRIEND
75740 Sigma-Aldrich

Oxalyldihydrazide

purum, ≥98.0% (N)

Synonym: Oxalic dihydrazide, Oxalylhydrazide

  • CAS Number 996-98-5

  • Linear Formula NH2NHCOCONHNH2

  • Molecular Weight 118.09

  •  Beilstein Registry Number 1072110

  •  EC Number 213-640-2

  •  MDL number MFCD00007608

  •  PubChem Substance ID 329766670

Purchase

Properties

Related Categories Building Blocks, Carbonyl Compounds, Chemical Synthesis, Hydrazides, Organic Building Blocks More...
InChI Key   SWRGUMCEJHQWEE-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (N)
mp   242-244 °C (dec.)(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
RO2840000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?