• USA Home
  • 75755 - Di(N-succinimidyl) oxalate

EMAIL THIS PAGE TO A FRIEND
75755 Sigma-Aldrich

Di(N-succinimidyl) oxalate

purum, ≥97.0% (T)

Synonym: N,N′-Disuccinimidyl oxalate, DSO

  • CAS Number 57296-03-4

  • Empirical Formula (Hill Notation) C10H8N2O8

  • Molecular Weight 284.18

  •  Beilstein Registry Number 4562450

  •  MDL number MFCD00011730

  •  PubChem Substance ID 329766683

Purchase

Properties

Related Categories Active Esters/Additives, Chemical Biology, Chemical Synthesis, Coupling, Peptide Chemistry,
InChI Key   OMAHFYGHUQSIEF-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (T)
mp   239 °C (dec.)(lit.)

Description

Other Notes

Reagent for preparing active esters of N-protected amino acids and for the preparation of protected dipeptides in good yields

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?