• USA Home
  • 75765 - Oxalyl chloride solution

EMAIL THIS PAGE TO A FRIEND
75765 Sigma-Aldrich

Oxalyl chloride solution

purum, ~2 M in methylene chloride

  • CAS Number 79-37-8

  • Linear Formula ClCOCOCl

  • Molecular Weight 126.93

  •  Beilstein Registry Number 1361988

  •  MDL number MFCD00000704

  •  PubChem Substance ID 329766692

Purchase

Properties

Related Categories Chemical Synthesis, Other Oxidizing Agents, Oxidizing Agents, Synthetic Reagents
InChI Key   CTSLXHKWHWQRSH-UHFFFAOYSA-N
grade   purum
concentration   ~2 M in methylene chloride

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Contact with water liberates toxic gas., Reacts violently with water.
RIDADR 
UN 2927 8(6.1) / PGII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

310670 Oxalyl chloride solution, 2.0 M in methylene chloride

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?