EMAIL THIS PAGE TO A FRIEND
75820 Sigma-Aldrich

2-Oxobutyric acid

purum, ≥97.0% (T)

Synonym: α-Ketobutyric acid, 2-Oxobutanoic acid, Propionylformic acid

  • CAS Number 600-18-0

  • Linear Formula CH3CH2COCOOH

  • Molecular Weight 102.09

  •  Beilstein Registry Number 1700514

  •  EC Number 209-986-9

  •  MDL number MFCD00004164

  •  PubChem Substance ID 329766722

Purchase

Properties

Related Categories Building Blocks, C1 to C5, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   TYEYBOSBBBHJIV-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (T)
mp   30-33 °C
solubility   H2O: 0.1 g/mL clear
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
179.6 °F
Flash Point(C) 
82 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

K401 2-Ketobutyric acid, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?