• USA Home
  • 75942 - Ethyl 2-oxo-4-phenylbutyrate

EMAIL THIS PAGE TO A FRIEND
75942 Sigma-Aldrich

Ethyl 2-oxo-4-phenylbutyrate

purum, ≥97.0% (GC)

  • CAS Number 64920-29-2

  • Linear Formula C6H5CH2CH2COCOOCH2CH3

  • Molecular Weight 206.24

  •  Beilstein Registry Number 2725083

  •  EC Number 265-276-9

  •  MDL number MFCD00037533

  •  PubChem Substance ID 329766784

Purchase

Properties

Related Categories Building Blocks, C12 to C63, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   STPXIOGYOLJXMZ-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.504(lit.)
  n20/D 1.504
bp   132 °C/2 mmHg(lit.)
density   1.091 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

375322 Ethyl 2-oxo-4-phenylbutyrate, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?