EMAIL THIS PAGE TO A FRIEND
75953 Sigma-Aldrich

Ethyl 3-oxovalerate

purum, ≥97.0% (GC)

  • CAS Number 4949-44-4

  • Linear Formula CH3CH2COCH2COOC2H5

  • Molecular Weight 144.17

  •  Beilstein Registry Number 970270

  •  EC Number 225-593-5

  •  MDL number MFCD00009317

  •  PubChem Substance ID 329766791

Purchase

Properties

Related Categories Building Blocks, C6 to C7, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   UDRCONFHWYGWFI-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.422(lit.)
  n20/D 1.424
bp   83-84 °C/12 mmHg(lit.)
density   1.012 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
172.4 °F
Flash Point(C) 
78 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?