• USA Home
  • 75962 - 3,3′-Oxydipropionitrile

EMAIL THIS PAGE TO A FRIEND
75962 Sigma-Aldrich

3,3′-Oxydipropionitrile

purum, ≥98.0% (GC)

Synonym: 2,2′-Dicyanodiethyl ether, Bis(2-cyanoethyl) ether

  • CAS Number 1656-48-0

  • Linear Formula O(CH2CH2CN)2

  • Molecular Weight 124.14

  •  Beilstein Registry Number 1746454

  •  EC Number 216-750-9

  •  MDL number MFCD00001960

Purchase

Properties

Related Categories Building Blocks, C6 to C7, Chemical Synthesis, Cyanides/Nitriles, Nitrogen Compounds,
InChI Key   BCGCCTGNWPKXJL-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.441(lit.)
  n20/D 1.441
bp   110-112 °C/0.5 mmHg(lit.)
density   1.043 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
UG4200000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

C91204 2-Cyanoethyl ether, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?