• USA Home
  • 76035 - Palladium standard for AAS

EMAIL THIS PAGE TO A FRIEND
76035 Sigma-Aldrich

Palladium standard for AAS

ready-to-use, in nitric acid

Synonym: Palladium(II) nitrate solution

  • Linear Formula Pd(NO3)2

  • Molecular Weight 230.43

  •  MDL number MFCD00011169

  •  PubChem Substance ID 24886925

Purchase

Properties

Related Categories AAS, Additional Standards, Analytical Standards, Analytical/Chromatography, Single Solution,
InChI Key   GPNDARIEYHPYAY-UHFFFAOYSA-N
shelf life   limited shelf life, expiry date on the label
concentration   in nitric acid
  1.000 g/L Pd
application(s)   ICP/AAS: suitable
format   single component solution

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Preparation Note

prepared with Pd(NO3)2 and HNO3 (~1 M)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3264 8 / PGIII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

78437 Palladium Standard for AAS, TraceCERT®, 1000 mg/L Pd in hydrochloric acid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?