• USA Home
  • 76044 - Palladium(II) acetate

EMAIL THIS PAGE TO A FRIEND
76044 Sigma-Aldrich

Palladium(II) acetate

purum, 47% Pd basis

Synonym: Pd(OAc)2

  • CAS Number 3375-31-3

  • Linear Formula Pd(OCOCH3)2

  • Molecular Weight 224.51

  •  Beilstein Registry Number 6086766

  •  EC Number 222-164-4

  •  MDL number MFCD00012453

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Homogeneous Pd Catalysts, Palladium
InChI Key   YJVFFLUZDVXJQI-UHFFFAOYSA-L
grade   purum
assay   47% Pd basis
mp   216.3-223.7 °C (dec.)

Description

Other Notes

Review

Same product — same quality — now Sigma-Aldrich brand

Application

Application Guide for Palladium Catalyzed Cross-Coupling Reactions

Price and Availability


Essence of Chemistry
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
AJ1900000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

205869 Palladium(II) acetate, reagent grade, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?