EMAIL THIS PAGE TO A FRIEND
76152 Sigma-Aldrich

Palmitoyl chloride

purum, ≥97.0% (GC)

Synonym: Hexadecanoyl chloride

  • CAS Number 112-67-4

  • Linear Formula CH3(CH2)14COCl

  • Molecular Weight 274.87

  •  Beilstein Registry Number 972409

  •  EC Number 203-996-7

  •  MDL number MFCD00000742

  •  PubChem Substance ID 329766554

Purchase

Properties

Related Categories Acid Halides, Building Blocks, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks More...
InChI Key   ARBOVOVUTSQWSS-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.452(lit.)
  n20/D 1.452
bp   88-90 °C/0.2 mmHg(lit.)
mp   11-13 °C(lit.)
density   0.907 g/mL at 20 °C
  0.906 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 3265 8 / PGII
WGK Germany 
1
Flash Point(F) 
320 °F
Flash Point(C) 
160 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?