• USA Home
  • 76190 - Pancreatin from porcine pancreas

EMAIL THIS PAGE TO A FRIEND
76190 Sigma-Aldrich

Pancreatin from porcine pancreas

powder, beige, amylase >100 U/mg

Synonym: Pancreatin from hog pancreas

Purchase

Properties

form   powder
color   beige
storage temp.   2-8°C

Description

Biochem/physiol Actions

Pancreatin contains enzymatic components including trypsin, amylase and lipase, ribonuclease, and protease, produced by the exocrine cells of the porcine pancreas. This combination of enzymes allows it to hydrolyze proteins, starch and fats. Pancreatin will convert not less than 25 times its weight of potato starch into soluble carbohydrates in 5 minutes in water at 40°C, will digest not less than 25 times its weight of casein in 60 minutes at pH 7.5 at 40°C and will release not less than microequivalents of acid per min per mg pancreatin from olive oil at pH 9.0 at 37°C.

Other Notes

In the digestion of casein and rapeseed; For the cleaning of reverse osmosis membranes; Regioselective acetylation of monosaccharide derivatives by pancreatin-catalyzed transesterification in organic solvents; Enantioselective lactonization of methyl 3,5-dihydroxyalkanoates

Sales restrictions may apply

Unit Definition

1 U corresponds to 1 USP-unit (USP XXI, p. 777) other enzyme activities: Lipase ≥8 U/mg [1 U corresponds to 1 USP-unit]; Protease ≥100 U/mg [1 U corresponds to 1 USP-unit]

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

WGK Germany 
1
RTECS 
RT9033000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P1750 Pancreatin from porcine pancreas, 4 × USP specifications

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?