EMAIL THIS PAGE TO A FRIEND
76243 Sigma-Aldrich

Paraffin wax

pellets, white

Purchase

Properties

Related Categories Core Bioreagents, Lipids and Related Products, Molecular Biology, Optimization Reagents, Protein Structural Analysis,
form   pellets
ign. residue   ≤0.05%
color   white
mp   43-95 °C
transition temp   solidification point 58-60 °C

Price and Availability


Laboratory Gloves

Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?