• USA Home
  • 76287 - Pectinase from Rhizopus sp.

EMAIL THIS PAGE TO A FRIEND
76287 Sigma-Aldrich

Pectinase from Rhizopus sp.

powder, slightly brown, ~200 U/g

Synonym: Macerozyme R-200, Poly-(1,4-α-D-galacturonide) glycanohydrolase, Polygalacturonase

Purchase

Description

Application

Used in plant protoplast preparation to digest cell wall prior to organelle isolation.

Unit Definition

1 U corresponds to the amount of enzyme which produces 1 μmol galacturonic acid per minute at pH 4 and 25°C (pectin from citrus peel, Sigma P3889, as substrate)

Price and Availability


Laboratory Gloves

Biomedical Applications
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?