EMAIL THIS PAGE TO A FRIEND
76588 Sigma-Aldrich

1-Phenylpentadecane

analytical standard

Synonym: Pentadecylbenzene

  • CAS Number 2131-18-2

  • Empirical Formula (Hill Notation) C21H36

  • Molecular Weight 288.51

  •  Beilstein Registry Number 1911130

  •  EC Number 218-352-0

  •  MDL number MFCD00015083

  •  PubChem Substance ID 329767190

Purchase

Properties

Related Categories Additional Standards, Alpha Sort, Analytical Standards, Analytical/Chromatography, Aromatics,
grade   analytical standard
InChI Key   JIRNEODMTPGRGV-UHFFFAOYSA-N
assay   ≥99.5% (GC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.482
bp   373 °C(lit.)
mp   22 °C(lit.)
density   0.855 g/mL at 20 °C(lit.)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?