• USA Home
  • 76746 - (S)-(−)-α-Methyl-2,3,4,5,6-pentafluorobenzyl alcohol

EMAIL THIS PAGE TO A FRIEND
76746 Sigma-Aldrich

(S)-(−)-α-Methyl-2,3,4,5,6-pentafluorobenzyl alcohol

for chiral derivatization, ≥99.0%

Synonym: (S)-(−)-1-(Pentafluorophenyl)ethanol

  • CAS Number 104371-20-2

  • Linear Formula CH3CH(C6F5)OH

  • Molecular Weight 212.12

  •  Beilstein Registry Number 4314037

  •  MDL number MFCD00077840

  •  PubChem Substance ID 329767314

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Chiral, Chiral Derivatization Reagents, Chiral GC Derivatization Reagents,
grade   for chiral derivatization
InChI Key   WYUNHWKTLDBPLE-REOHCLBHSA-N
assay   ≥99.0% (sum of enantiomers, GC)
  ≥99.0%
optical activity   [α]20/D −7.0±0.5°, c = 1% in pentane
optical purity   enantiomeric ratio: ≥99.5:0.5 (GC)
mp   40-43 °C
  41-42 °C(lit.)

Description

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
188.6 °F
Flash Point(C) 
87 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?