• USA Home
  • 76765 - 2,3,4,5,6-Pentafluorostyrene

EMAIL THIS PAGE TO A FRIEND
76765 Sigma-Aldrich

2,3,4,5,6-Pentafluorostyrene

purum, ≥98.0% (GC)

  • CAS Number 653-34-9

  • Linear Formula C6F5CH=CH2

  • Molecular Weight 194.10

  •  Beilstein Registry Number 1874390

  •  EC Number 211-500-5

  •  MDL number MFCD00000300

  •  PubChem Substance ID 24887054

Purchase

Properties

Related Categories Alkenyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
InChI Key   LVJZCPNIJXVIAT-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
impurities   ≤0.1% water
refractive index   n20/D 1.446(lit.)
  n20/D 1.446
bp   62-63 °C/50 mmHg(lit.)
density   1.406 g/mL at 25 °C(lit.)
shipped in   wet ice
storage temp.   −20°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
93.2 °F
Flash Point(C) 
34 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

196916 2,3,4,5,6-Pentafluorostyrene, 99%, contains 0.1% p-tert-butylcatechol as inhibitor

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?