• USA Home
  • 76828 - N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine

EMAIL THIS PAGE TO A FRIEND
76828 Sigma-Aldrich

N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine

purum, ≥98.0% (GC)

Synonym: PMDETA, PMDTA

  • CAS Number 3030-47-5

  • Linear Formula [(CH3)2NCH2CH2]2NCH3

  • Molecular Weight 173.30

  •  Beilstein Registry Number 1741396

  •  EC Number 221-201-1

  •  MDL number MFCD00014876

  •  PubChem Substance ID 24887060

Purchase

Properties

vapor pressure   0.23 mmHg ( 20 °C)
InChI Key   UKODFQOELJFMII-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
autoignition temp.   311 °F
expl. lim.   5.6 %
impurities   ≤0.5% water
refractive index   n20/D 1.442(lit.)
  n20/D 1.443
bp   198 °C(lit.)
mp   −20 °C(lit.)
density   0.83 g/mL at 25 °C(lit.)

Description

Other Notes

Catalyst component for Ni(II)-mediated alkyne carboxylation

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2735PSN2 8 / PGII
WGK Germany 
2
Flash Point(F) 
170.6 °F
Flash Point(C) 
77 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

369497 N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?