• USA Home
  • 76857 - Benzalkonium chloride solution

EMAIL THIS PAGE TO A FRIEND
76857 Sigma-Aldrich

Benzalkonium chloride solution

PharmaGrade, EP, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production

Synonym: Alkylbenzyldimethylammonium chloride, Benzalkonii chloridi solution

Purchase

Properties

Related Categories Ammonium Salts, Biochemicals and Reagents, Chemical Synthesis, Detergents, Detergents A to M,
Quality Level   GMP-COMPENDIA
biological source   synthetic
grade   EP
  Ph Eur
  USP/NF
  USP
product line   PharmaGrade
form   liquid
pharmacopeia   testing & handling conforms to Pharmacopeia
concentration   50% in H2O
impurities   amine and amine salt, complies
  ≤0.05% impurity C : benzylchloride (HPLC)
  ≤0.1% sulfated ash
  ≤0.15% impurity B : benzaldehyde (HPLC)
  ≤0.5% impurity A : benzyl alcohol (HPLC)
suitability   complies for acidity or alkalinity

Description

Analysis Note

Chromatography (HPLC)

relative molecular mass
• determine

ratio of alkyl components
• C12 Homologue ≥40.0 % w/w
• C14 Homologue ≥20.0 % w/w
• C12 + C14 Homologue ≥70.0 % w/w

Identification
• Ph. Eur. identification B and NF identification D (HPLC) :tests corresponds to requirements
• Ph. Eur. identification E and NF identification A and B :tests corresponds to requirements
• NF identification C : corresponds to requirements

appearance of solution (Ph. Eur.)
• clear solution (≤3.5 NTU) not more intensely colored than reference solution G6

Ph.Eur. and NF assay (titration)
• 475 g/L - 525 g/L (corresponds to 95.0 - 105.0%)

General description

PharmaGrade products are manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production. For more information on the PharmaGrade Program, visit sigma-aldrich.com/safc-pharmagrade.

Linkage

Enhanced Quality Product Documentation on PharmaGrade aq. Benzalkonium Chloride Solution

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN3265 - class 8 - PG 2 - EHS - Corrosive liquid, acidic, organic, n.o.s., HI: all
WGK Germany 
3
Protocols & Articles

Related Content

Enhanced Quality Program [VIDEO]

The Enhanced Quality Program is a quality management system that is used to differentiate products based on the required level of transparency, control and documentation support determined by the nee...

One Quality Policy [VIDEO]

Sigma-Aldrich maintains one quality policy on a global level to provide consistency in vision, maintained across all manufacturing and distribution sites so that the company on the whole is adhering ...

Quality Management Systems [VIDEO]

See how we combine the use of various Quality Management Systems to manage manufacturing and supply chain for our products, as well as ensure data integrity for information provided to customers via ...

Risk Management [VIDEO]

Our Quality Policy means that we are able to provide certain information to end customers based on a product’s Enhanced Quality level, and that information is limited to what we are able to guarantee...

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?