EMAIL THIS PAGE TO A FRIEND
76874 Sigma-Aldrich

Pentane

purum, ≥95.0% (GC)

  • CAS Number 109-66-0

  • Linear Formula CH3(CH2)3CH3

  • Molecular Weight 72.15

  •  Beilstein Registry Number 969132

  •  EC Number 203-692-4

  •  MDL number MFCD00009498

Purchase

Properties

vapor density   2.48 (vs air)
InChI Key   OFBQJSOFQDEBGM-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
form   liquid
autoignition temp.   500 °F
expl. lim.   ~8.3 %
refractive index   n20/D 1.358(lit.)
  n20/D 1.358
bp   35-36 °C(lit.)
mp   −130 °C(lit.)
solubility   ethanol: soluble(lit.)
density   0.626 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN1265 - class 3 - PG 2 - Pentanes
WGK Germany 
2
RTECS 
RZ9450000
Flash Point(F) 
-56.2 °F
Flash Point(C) 
-49 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?