EMAIL THIS PAGE TO A FRIEND
76880 Sigma-Aldrich

Pentane Fraction

purum

Purchase

Properties

InChI Key   OFBQJSOFQDEBGM-UHFFFAOYSA-N
grade   purum
refractive index   n20/D 1.358
bp   32-37 °C
density   0.625 g/mL at 20 °C

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN1265 - class 3 - PG 2 - Pentanes
WGK Germany 
2
Flash Point(F) 
-40 °F
Flash Point(C) 
-40 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?