EMAIL THIS PAGE TO A FRIEND
76942 Sigma-Aldrich

(±)-2-Pentanol

purum, ≥98.0% (GC)

Synonym: sec-Pentyl alcohol, Methyl propyl carbinol

  • CAS Number 6032-29-7

  • Linear Formula CH3CH2CH2CH(OH)CH3

  • Molecular Weight 88.15

  •  Beilstein Registry Number 1718819

  •  EC Number 227-907-6

  •  MDL number MFCD00004579

  •  PubChem Substance ID 329767442

Purchase

Properties

Related Categories Alcohols, Building Blocks, C2 to C6, Chemical Synthesis, Organic Building Blocks,
vapor density   3 (vs air)
InChI Key   JYVLIDXNZAXMDK-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
autoignition temp.   649 °F
expl. lim.   9 %
refractive index   n20/D 1.406(lit.)
  n20/D 1.407
bp   118-119 °C(lit.)
density   0.812 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN1105 - class 3 - PG 3 - Pentanols
WGK Germany 
2
RTECS 
SA4900000
Flash Point(F) 
93.2 °F
Flash Point(C) 
34 °C
Protocols & Articles

Articles

GC Analysis of Alcohols on PAG

From our library of Articles, Sigma-Aldrich presents GC Analysis of Alcohols on PAG
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

GC Analysis of Fermentation Compounds on a Packed Column (6.6% Carbowax® 20M on 80/120 °Carbopack™ B AW)

From our library of Articles, Sigma-Aldrich presents GC Analysis of Fermentation Compounds on a Packed Column (6.6% Carbowax® 20M on 80/120 °Carbopack™ B AW)
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?