• USA Home
  • 76953 - Sodium 1-pentanesulfonate monohydrate

EMAIL THIS PAGE TO A FRIEND
76953 Sigma-Aldrich

Sodium 1-pentanesulfonate monohydrate

≥99.0% (T)

Synonym: 1-Pentanesulfonic acid sodium salt

  • CAS Number 207605-40-1

  • Linear Formula CH3(CH2)4SO3Na · H2O

  • Molecular Weight 192.21

  •  Beilstein Registry Number 3725147

  •  EC Number 245-208-4

  •  MDL number MFCD00149548

  •  PubChem Substance ID 24887089

Purchase

Properties

Related Categories Alkyl Sulfonates, Anionic Detergents, Biochemicals and Reagents, Building Blocks, Chemical Synthesis,
description   anionic
InChI Key   FPQYXAFKHLSWTI-UHFFFAOYSA-M
assay   ≥99.0% (T)
solubility   H2O: soluble0.1 g/mL, clear, colorless

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?