EMAIL THIS PAGE TO A FRIEND
77020 Sigma-Aldrich

4-Penten-1-ol

purum, ≥97.0% (GC)

Synonym: 2-Allylethyl alcohol

  • CAS Number 821-09-0

  • Linear Formula CH2=CH(CH2)3OH

  • Molecular Weight 86.13

  •  Beilstein Registry Number 1560163

  •  EC Number 212-473-2

  •  MDL number MFCD00002975

  •  PubChem Substance ID 329767457

Purchase

Properties

Related Categories Acyclic, Alkenes, Biochemicals and Reagents, Building Blocks, Chemical Synthesis,
InChI Key   LQAVWYMTUMSFBE-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.429(lit.)
  n20/D 1.429
bp   134-137 °C(lit.)
density   0.834 g/mL at 25 °C(lit.)

Description

Other Notes

Preparation of pentenyl ribosides, protected carbohydrates for nucleoside synthesis

Price and Availability


The Future of Bioimaging
Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1987C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
118.4 °F
Flash Point(C) 
48 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

111279 4-Penten-1-ol, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?