EMAIL THIS PAGE TO A FRIEND
77040 Sigma-Aldrich

4-Penten-2-ol

purum, ≥98.0% (GC)

Synonym: Allyl methyl carbinol

  • CAS Number 625-31-0

  • Linear Formula H2C=CHCH2CH(OH)CH3

  • Molecular Weight 86.13

  •  Beilstein Registry Number 1839744

  •  EC Number 210-887-8

  •  MDL number MFCD00004556

  •  PubChem Substance ID 329767458

Purchase

Properties

Related Categories Acyclic, Alkenes, Building Blocks, Chemical Synthesis, Organic Building Blocks More...
InChI Key   ZHZCYWWNFQUZOR-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.424(lit.)
  n20/D 1.424
bp   115-116 °C(lit.)
density   0.837 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1987C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
86 °F
Flash Point(C) 
30 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?