EMAIL THIS PAGE TO A FRIEND
77055 Sigma-Aldrich

4-Pentynoic acid

purum, ≥97% (GC)

Synonym: Propargylacetic acid

Purchase

Properties

Related Categories Building Blocks, C1 to C5, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   MLBYLEUJXUBIJJ-UHFFFAOYSA-N
grade   purum
assay   ≥97% (GC)
bp   110 °C/30 mmHg(lit.)
mp   54-57 °C(lit.)
  54-57 °C
storage temp.   2-8°C

Description

Other Notes

Versatile building block for the synthesis of 4-alkynoic acids by C-alkylation

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3
RTECS 
SC4751000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

232211 4-Pentynoic acid, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?