EMAIL THIS PAGE TO A FRIEND
77080 Sigma-Aldrich

Pentylbenzene

purum

Synonym: 1-Phenylpentane

  • CAS Number 538-68-1

  • Linear Formula C6H5(CH2)4CH3

  • Molecular Weight 148.24

  •  Beilstein Registry Number 1904359

  •  EC Number 208-701-5

  •  MDL number MFCD00009502

  •  PubChem Substance ID 329767464

Purchase

Properties

Related Categories Arenes, Building Blocks, Chemical Synthesis, Organic Building Blocks
InChI Key   PWATWSYOIIXYMA-UHFFFAOYSA-N
grade   purum
refractive index   n20/D 1.488(lit.)
  n20/D 1.489
bp   205 °C(lit.)
mp   −75 °C(lit.)
density   0.863 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN3082 - class 9 - PG 3 - MP - DOT NA1993 - Environmentally hazardous substances, liquid, n.o.s. HI: all (not BR)
WGK Germany 
3
Flash Point(F) 
150.8 °F
Flash Point(C) 
66 °C
Protocols & Articles

Articles

HPLC Analysis of Pentylbenzene and o-Terphenyl on Ascentis® Express Biphenyl

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Pentylbenzene and o-Terphenyl on Ascentis® Express Biphenyl
Keywords: Chromatography, High performance liquid chromatography, Pharmaceutical, Separation

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

113174 Pentylbenzene, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?