• USA Home
  • 77265 - Pentafluoroiodoethane

EMAIL THIS PAGE TO A FRIEND
77265 Sigma-Aldrich

Pentafluoroiodoethane

purum, ≥97.0% (GC)

Synonym: Iodopentafluoroethane, Perfluoroethyl iodide

  • CAS Number 354-64-3

  • Linear Formula CF3CF2I

  • Molecular Weight 245.92

  •  Beilstein Registry Number 1740331

  •  EC Number 206-566-7

  •  MDL number MFCD00039400

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Chemical Synthesis, Derivatization Reagents, Derivatization Reagents GC,
vapor pressure   738.9 mmHg ( 10 °C)
InChI Key   UXPOJVLZTPGWFX-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.339(lit.)
bp   12-13 °C(lit.)
density   2.085 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Risk of explosion if heated under confinement.
RIDADR 
UN 1956 2.2
WGK Germany 
3
RTECS 
KI6397500
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

331015 Pentafluoroiodoethane, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?