EMAIL THIS PAGE TO A FRIEND
77275 Sigma-Aldrich

Perfluorokerosene

for mass spectrometry, high boiling, high viscosity

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Mass Spectrometry, Mass Spectrometry (MS), Matrices for MALDI Analysis,
grade   for mass spectrometry
refractive index   n20/D 1.330
bp   ≥300 °C
  >300 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?