EMAIL THIS PAGE TO A FRIEND
77277 Sigma-Aldrich

Perfluorokerosene

for mass spectrometry, low boiling

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Mass Spectrometry (MS), Reagents for Mass Spectrometry (MS), Spectroscopy More...
grade   for mass spectrometry
refractive index   n20/D 1.300
bp   >300 °C(lit.)
  70-110 °C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
114.8 °F
Flash Point(C) 
46 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?