• USA Home
  • 77288 - Heptadecafluorooctanesulfonic acid tetraethylammonium salt

EMAIL THIS PAGE TO A FRIEND
77288 Sigma-Aldrich

Heptadecafluorooctanesulfonic acid tetraethylammonium salt

purum, ≥98.0% (T)

Synonym: Perfluorooctanesulfonic acid tetraethylammonium salt

  • CAS Number 56773-42-3

  • Linear Formula CF3(CF2)6CF2SO3N(C2H5)4

  • Molecular Weight 629.37

  •  Beilstein Registry Number 6470030

  •  EC Number 260-375-3

  •  MDL number MFCD00066497

  •  PubChem Substance ID 329767582

Purchase

Properties

Related Categories Ammonium Salts, Chemical Synthesis, Phase Transfer Catalysts, Synthetic Reagents
InChI Key   JHDXAQHGAJXNBY-UHFFFAOYSA-M
grade   purum
assay   ≥98.0% (T)
mp   183-190 °C
  184-190 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
2
RTECS 
KH3154250

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

365289 Heptadecafluorooctanesulfonic acid tetraethylammonium salt, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?