EMAIL THIS PAGE TO A FRIEND
77382 Sigma-Aldrich

Petroleum ether

B&J Brand (product of Burdick & Jackson), bp 30-60 °C

Synonym: Petroleum benzin

Purchase

Properties

grade   B&J Brand (product of Burdick & Jackson)
bp   30-60 °C

Description

Other Notes

Sales restrictions may apply

Same product — same quality — now Sigma-Aldrich brand

General description

Special brochure (85475) available at cost price.

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Precautionary statements 
RIDADR 
UN1268 - class 3 - PG 2 - EHS - VP > 110kPa - Petroleum distillates, n.o.s., HI: S
WGK Germany 
2
Flash Point(F) 
-40 °F
Flash Point(C) 
-40 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?