77399 Sigma-Aldrich

Petroleum ether

bp 30-40 °C, low boiling point hydrogen treated naphtha, puriss. p.a.

DOWNLOAD MSDS (PDF)

Synonym: Petroleum benzin

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, O-P, Puriss p.a., Puriss p.a. More...
grade   puriss. p.a.
dist. (ASTM D86-87)   ≥95%between 30 and 40°C
impurities   substances darkened by H2SO4, in accordance
  ≤0.002% free acid (as CH3COOH)
  ≤0.005% S-compounds (as S)
  ≤0.01% benzene (GC)
  ≤0.02% water
evapn. residue   ≤0.001%
refractive index   n20/D 1.357
bp   30-40 °C
density   0.63 g/mL at 20 °C
cation traces   Al: ≤0.5 mg/kg
  B: ≤0.02 mg/kg
  Ba: ≤0.1 mg/kg
  Bi: ≤0.1 mg/kg
  Ca: ≤0.5 mg/kg
  Cd: ≤0.05 mg/kg
  Co: ≤0.02 mg/kg
  Cr: ≤0.02 mg/kg
  Cu: ≤0.02 mg/kg
  Fe: ≤0.1 mg/kg
  K: ≤0.5 mg/kg
  Li: ≤0.1 mg/kg
  Mg: ≤0.1 mg/kg
  Mn: ≤0.02 mg/kg
  Mo: ≤0.1 mg/kg
  Na: ≤0.5 mg/kg
  Ni: ≤0.02 mg/kg
  Pb: ≤0.02 mg/kg
  Sn: ≤0.1 mg/kg
  Sr: ≤0.1 mg/kg
  Zn: ≤0.1 mg/kg

Price and Availability

Customers Also Viewed

Diethyl ether

contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%

DIN 51635, bp 65-95 °C

anhydrous

puriss. p.a., low boiling point hydrogen treated naphtha, bp 30-50 °C

puriss. p.a., ACS reagent, bp 40-60 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?