• USA Home
  • 77415 - Phalloidin – fluorescein isothiocyanate conjugate from Amanita phalloides

EMAIL THIS PAGE TO A FRIEND
77415 Sigma-Aldrich

Phalloidin – fluorescein isothiocyanate conjugate from Amanita phalloides

for fluorescence, mixture of isomers

Synonym: FITC - Phalloidin conjugate

Purchase

Properties

Related Categories Albumin and other protein conjugates, Biochemicals and Reagents, Fluorescent Conjugates, Fluorescent Conjugates for Microscopy, Fluorescent Probes, Labels, Particles and Stains More...
grade   for fluorescence
extent of labeling   0.001-0.008 mol
shipped in   wet ice
storage temp.   −20°C

Description

Packaging

package with 100 μg

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2811 6.1/PG 2
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

P5282 Phalloidin, Fluorescein Isothiocyanate Labeled, Phalloidin from Amanita phalloides

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?