• USA Home
  • 77418 - Phalloidin – tetramethylrhodamine B isothiocyanate conjugate from Amanita phalloides

EMAIL THIS PAGE TO A FRIEND
77418 Sigma-Aldrich

Phalloidin – tetramethylrhodamine B isothiocyanate conjugate from Amanita phalloides

for fluorescence, mixture of isomers

Synonym: Phalloidin-TRITC labeled, Tetramethylrhodamine B isothiocyanate Phalloidin Conjugate

Purchase

Properties

Related Categories Albumin and other protein conjugates, Biochemicals and Reagents, Fluorescent Conjugates, Fluorescent Conjugates for Microscopy, Fluorescent Labels,
grade   for fluorescence
solubility   methanol: soluble1 mg/mL, clear, bright pink
shipped in   wet ice
storage temp.   −20°C

Description

Frequently Asked Questions

Frequently Asked Questions are available for this Product.

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Packaging

package with 100 μg

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol, solution
WGK Germany 
2
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?